Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, 25,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 150,000
Supelco
Dextran, analytical standard, 5,000
Supelco
Dextran aus Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 670,000
Supelco
Dextran, analytical standard, suitable for gel permeation chromatography (GPC), Set Mp 1,000-400,000
Supelco
Dextran, analytical standard, 670,000
Supelco
Dextran, analytical standard, 150,000
Supelco
Dextran, analytical standard, 1,000
Supelco
Dextran, analytical standard, 270,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran, analytical standard, 12,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran -Lösung aus Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, Mr ~60,000
Supelco
Dextran, analytical standard, 410,000
Sigma-Aldrich
Dextran, enzymatic synth.
Supelco
Dextran, analytical standard, 50,000
Supelco
Dextran aus Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 5,000
Supelco
Dextran, analytical standard, 80,000
Supelco
Dextran aus Leuconostoc mesenteroides, analytical standard, Mw 1,000
Supelco
Dextran aus Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 25,000