Merck
  • The role of clobetasol propionate emollient 0.05% in the treatment of patients with dry, scaly, corticosteroid-responsive dermatoses.
The role of clobetasol propionate emollient 0.05% in the treatment of patients with dry, scaly, corticosteroid-responsive dermatoses.
Clinical therapeutics (1998-04-02)
M L Gordon
ZUSAMMENFASSUNG
The use of topical corticosteroids has significantly enhanced the treatment of patients with dermatoses such as psoriasis and eczema. In particular, group I high-potency corticosteroids such as clobetasol propionate have proved safe and effective for limited-course treatment of inflammatory and pruritic manifestations of moderate-to-severe corticosteroid-responsive dermatoses. At the same time, much effort has gone into devising more effective strategies for addressing the dry skin conditions associated with various dermatologic disorders. An emollient added to a steroid, although not itself an active ingredient, can help restore the normal moisturizing process of the skin; this may be particularly important in soothing the discomfort of the dry skin conditions often encountered in moderate-to-severe dermatoses. In addition, the degree of epidermal hydration can affect the penetration of steroids into the skin. Therefore, successful outcomes in the treatment of patients with corticosteroid-responsive dermatoses may involve more than use of an effective topical steroid. This article examines a currently available cream formulation of 0.05% clobetasol propionate containing moisturizers--emollients, dimethicone, and a humectant--that may contribute to improved moisture content in treated skin. A review of recent studies shows that clobetasol propionate emollient cream is well tolerated and effective in courses of up to 4 weeks for the treatment of patients with psoriasis or atopic dermatitis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Clobetasolpropionat, United States Pharmacopeia (USP) Reference Standard
Supelco
Clobetasolpropionat, analytical standard
Clobetasolpropionat, European Pharmacopoeia (EP) Reference Standard