Merck
Thymopentin and splenopentin as immunomodulators. Current status.
Immunologic research (1998-06-25)
V K Singh, S Biswas, K B Mathur, W Haq, S K Garg, S S Agarwal
ZUSAMMENFASSUNG
Splenopentin (SP-5, Arg-Lys-Glu-Val-Tyr) and thymopentin (TP-5, Arg-Lys-Asp-Val-Tyr) are synthetic immunomodulating peptides corresponding to the region 32-34 of a splenic product called splenin (SP) and the thymic hormone thymopoietin (TP), respectively. TP was originally isolated as a 5-kDa (49-amino acids) protein from bovine thymus while studying effects of the thymic extracts on neuromuscular transmission and was subsequently observed to affect T cell differentiation and function. TP I and II are two closely related polypeptides isolated from bovine thymus. A radioimmunoassay for TP revealed a crossreaction with a product found in spleen and lymph node. This product, named splenin, differs from TP only in position 34, aspartic acid for bovine TP and glutamic acid for bovine splenin and it was called TP III as well. Synthetic pentapeptides (TP-5) and (SP-5), reproduce the biological activities of TP and SP, respectively. It is now evident that various forms of TPs were created by proteolytic cleavage of larger proteins during isolation. cDNA clones have been isolated for three alternatively spliced mRNAs that encodes three distinct human T cell TPs. The immunomodulatory properties of TP, SP, TP-5, SP-5 and some of their synthetic analogs reported in the literature have been briefly reviewed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thymopentin, ≥97% (HPLC)