Merck
  • Saving periodontally "hopeless teeth" using calcium sulfate and demineralized freeze-dried bone allograft.
Saving periodontally "hopeless teeth" using calcium sulfate and demineralized freeze-dried bone allograft.
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995) (1998-05-20)
D Anson
ZUSAMMENFASSUNG
As the techniques of guided tissue regeneration (GTR) and implant placement evolve, the decision of when a tooth should be saved using GTR, or extracted and an implant placed, becomes much more complicated. This article discusses treating teeth with GTR that have traditionally had hopeless prognoses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Drierit, with indicator, 8 mesh
Sigma-Aldrich
Drierit, with indicator, 10-20 mesh
Sigma-Aldrich
Calciumsulfat, −325 mesh, 99%
Sigma-Aldrich
Drierit, with indicator, 4 mesh
Sigma-Aldrich
Drierit, with indicator, 6 mesh
Sigma-Aldrich
Calciumsulfat, ≥99.99% trace metals basis