Merck
  • How is chemical interesterification initiated: Nucleophilic substitution or a-proton abstraction?.
How is chemical interesterification initiated: Nucleophilic substitution or a-proton abstraction?.
Journal of the American Oil Chemists' Society (2004)
Liu L.