Merck
  • Quinone Imines with a Fused Azine Ring: I. Synthesis and Hydrochlorination of 5-(p-Tolylsulfonylimino) quinolin-8-one.
Quinone Imines with a Fused Azine Ring: I. Synthesis and Hydrochlorination of 5-(p-Tolylsulfonylimino) quinolin-8-one.
Russ. J. Org. Chem. (2004)
Belov AV and Nichvoloda VM.