Merck
  • Production of LYZL6, a novel human c-type lysozyme, in recombinant Pichia pastoris employing high cell density fed-batch fermentation.
Production of LYZL6, a novel human c-type lysozyme, in recombinant Pichia pastoris employing high cell density fed-batch fermentation.
Journal of bioscience and bioengineering (2014-04-22)
Xiaoyu Zhou, Ying Yu, Jianjun Tao, Long Yu
ZUSAMMENFASSUNG
Lysozyme acts as an important defensive factor in innate immunity due to its well-recognized bacteriolytic activity. Here we describe the production and performance of human lysozyme-like 6 (LYZL6), a novel human c-type lysozyme homolog. A synthetic codon-optimized cDNA encoding the intact amino acid sequence of LYZL6 was cloned and expressed in Pichia pastoris SMD1168. Bioactive LYZL6 was successfully produced as a single major secreted protein with a molecular weight of 15 kDa, and exhibited bacteriolytic activity against Micrococcus lysodeikticus. The expression conditions were optimized, and the highest expression level of LYZL6 occurred when the recombinant strain was induced with 1.5% methanol under pH 4.5 at 24°C for 96 h. When high cell density fermentation of the recombinant P. pastoris was performed using a fed-batch strategy for totally 125 h in a 30 L fermenter, the dry cell weight and the extracellular lysozyme activity were increased to 116.3 g/L and 2340 U/mL, respectively. The LYZL6 protein concentration was 331 mg/L of fermentation supernatant, and the specific activity of LYZL6 towards M. lysodeikticus was 7069 U/mg. Therefore, we proved that LYZL6 is an antibacterial protein, suggesting a potential application of LYZL6 as an antimicrobial agent, and Pichia expression system for LYZL6 was successful and industrially promising.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.