Merck
Tissue-specific subunit of the mouse cytosolic chaperonin-containing TCP-1.
FEBS letters (1997-01-27)
H Kubota, G M Hynes, S M Kerr, K R Willison
ZUSAMMENFASSUNG
We have cloned a novel Tcp-1-related mouse testis cDNA encoding a polypeptide of 531 amino acids which shares 81.2% identity with the zeta subunit of the mouse cytosolic chaperonin-containing TCP-1 (CCT). Immunoblot analysis of mouse testis CCT subunits separated by 2-dimensional gel electrophoresis indicates that this novel gene, Cctz-2, encodes a CCT subunit of Mr 57 000 and pI 7.1. Cctz-2 mRNA is detected only in testis whereas the other Cctz gene, Cctz-1, is expressed in all tissues investigated. The CCTzeta-2 subunit may have specific functions in the folding of testicular proteins and for interactions with testicular molecular chaperones.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-CCT6B antibody produced in rabbit, affinity isolated antibody