Merck
  • Supercritical fluid extraction as a means of cleaning and desorbing common air sampling sorbents.
Supercritical fluid extraction as a means of cleaning and desorbing common air sampling sorbents.
Chemosphere (1995)
Bowyer JR and Joachim DP.