Merck
  • Keratinolytic activity from the broth of a feather-degrading thermophillic Streptomyces thermoviolaceus strain SD8.
Keratinolytic activity from the broth of a feather-degrading thermophillic Streptomyces thermoviolaceus strain SD8.
Letters in Applied Microbiology (1999)
Chitte RR, Nalawade VK, Dey S.