Merck
The role of COX-2 in intestinal cancer.
Annals of the New York Academy of Sciences (2000-02-11)
C Williams, R L Shattuck-Brandt, R N DuBois
ZUSAMMENFASSUNG
Cyclooxygenase (COX), the key regulatory enzyme for prostaglandin synthesis is transcribed from two distinct genes. COX-1 is expressed constitutively in most tissues, and COX-2 is induced by a wide variety of stimuli and was initially identified as an immediate-early growth response gene. In addition, COX-2 expression is markedly increased in 85-90% of human colorectal adenocarcinomas, whereas COX-1 levels remain unchanged. Several epidemiological studies have reported a 40-50% reduction in the risk of developing colorectal cancer in persons who chronically take such nonsteroidal anti-inflammatory drugs (NSAIDs) as aspirin, which are classic inhibitors of cyclooxygenase. Genetic evidence also supports a role for COX-2, since mice null for COX-2 have an 86% reduction in tumor multiplicity in a background containing a mutated APC allele. These results strongly suggest that COX-2 contributes to the development of intestinal tumors and that inhibition of COX is chemo-preventative.