Merck
  • Supersonic jet fluorescence spectroscopy of 1, 3-benzodioxole: A non-planar (C< sub> 2</sub>) structure in S0.
Supersonic jet fluorescence spectroscopy of 1, 3-benzodioxole: A non-planar (C&lt; sub&gt; 2&lt;/sub&gt;) structure in S<SUB>0</SUB>.
Chemical Physics Letters (1989)
Hassan KM and Hollas JM.