Merck
  • Evaluation of Tenax-GR adsorbent for the passive sampling of volatile organic compounds at low concentrations.
Evaluation of Tenax-GR adsorbent for the passive sampling of volatile organic compounds at low concentrations.
Atmos. Environ., Part A (1993)
Cao X and Hewitt N.