Merck
  • Rapid and sensitive determination of phenylurea herbicides in water in the presence of their anilines by extraction with a Carbopack cartridge followed by liquid chromatography.
Rapid and sensitive determination of phenylurea herbicides in water in the presence of their anilines by extraction with a Carbopack cartridge followed by liquid chromatography.
Journal of Chromatography A (1991)
di Corcia A and Marchetti M.