Merck
  • The Aβ-clearance protein transthyretin, like neprilysin, is epigenetically regulated by the amyloid precursor protein intracellular domain.
The Aβ-clearance protein transthyretin, like neprilysin, is epigenetically regulated by the amyloid precursor protein intracellular domain.
Journal of neurochemistry (2014-02-18)
Caroline Kerridge, Nikolai D Belyaev, Natalia N Nalivaeva, Anthony J Turner
ZUSAMMENFASSUNG
Proteolytic cleavage of the amyloid precursor protein (APP) by the successive actions of β- and γ-secretases generates several biologically active metabolites including the amyloid β-peptide (Aβ) and the APP intracellular domain (AICD). By analogy with the Notch signalling pathway, AICD has been proposed to play a role in transcriptional regulation. Among the cohort of genes regulated by AICD is the Aβ-degrading enzyme neprilysin (NEP). AICD binds to the NEP promoter causing transcriptional activation by competitive replacement with histone deacetylases (HDACs) leading to increased levels of NEP activity and hence increased Aβ clearance. We now show that the Aβ-clearance protein transthyretin (TTR) is also epigenetically up-regulated by AICD. Like NEP regulation, AICD derived specifically from the neuronal APP isoform, APP695 , binds directly to the TTR promoter displacing HDAC1 and HDAC3. Cell treatment with the tyrosine kinase inhibitor Gleevec (imatinib) or with the alkalizing agent NH4 Cl causes an accumulation of 'functional' AICD capable of up-regulating both TTR and NEP, leading to a reduction in total cellular Aβ levels. Pharmacological regulation of both NEP and TTR might represent a viable therapeutic target in Alzheimer's disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumchlorid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammoniumchlorid, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammoniumchlorid, 99.998% trace metals basis
Supelco
Ammoniumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Ammoniumchlorid, 99.99% trace metals basis
Sigma-Aldrich
Ammoniumchlorid, BioUltra, ≥99.5% (AT)
Supelco
Ammonium-Ionenlösung für ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Ammoniumchlorid, tested according to Ph. Eur.