Merck
  • Transapical neochord implantation: is tension of artificial chordae tendineae dependent on the insertion site?
Transapical neochord implantation: is tension of artificial chordae tendineae dependent on the insertion site?
The Journal of thoracic and cardiovascular surgery (2013-09-18)
Henrik Jensen, Morten O Jensen, Farhad Waziri, Jesper L Honge, Erik Sloth, Morten Fenger-Gron, Sten L Nielsen
ZUSAMMENFASSUNG
Transapical chordae tendineae replacement is a promising new approach for mitral leaflet prolapse. However, animal studies have raised concerns that the tension of the transapically fixated artificial neochordae might be greater than the tension in the neochordae attached to papillary muscle tips, thereby reducing repair durability. In eight 80-kg pigs, the primary anterior leaflet chordae were replaced by a 5-0 polytetrafluoroethylene neochord using a miniature in-line force transducer. The neochord was attached first to the anterior papillary muscle and, on a second cardiac bypass, transapically to the left ventricle apex. Occlusion of the inferior vena cava was performed to examine the effect of left ventricle pressure changes on neochord tension to adjust the crude data to 95 mm Hg. The maximum slope of the chordal tension curve was calculated to compare curve patterns. The data are presented as the mean ± standard deviation. The following tension was measured in the neochordae during papillary muscle and transapical fixation, respectively: peak tension (crude, 0.39 ± 0.32 vs 0.50 ± 0.25 N, P = .17; adjusted, 0.41 ± 0.30 vs 0.46 ± 0.27 N, P = .22), mid-systolic tension (crude, 0.19 ± 0.12 vs 0.19 ± 0.15 N, P = .96; adjusted, 0.28 ± 0.16 vs 0.19 ± 0.11 N, P = .12). There was a significantly lower maximum slope (dF/dtmax) of the neochord tension curves after papillary muscle fixation compared with transapical fixation (7.4 ± 6.9 vs 10.3 ± 7.7 N/s, P = .028). Overall, the chordal insertion site had little influence on the tension in the artificial neochordae compared with the interindividual variation. However, abnormal tension fluctuations in the transapically fixated neochordae might predispose to leaflet tears and early repair failure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polytetrafluorethylen, 60 wt % dispersion in H2O
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), beads
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, ≥350 μm particle size