Merck
  • Study the effect of formulation variables on drug release from hydrophilic matrix tablets of milnacipran and prediction of in-vivo plasma profile.
Study the effect of formulation variables on drug release from hydrophilic matrix tablets of milnacipran and prediction of in-vivo plasma profile.
Pharmaceutical development and technology (2013-08-13)
Gautam Singhvi, Abhishek Shah, Nilesh Yadav, Ranendra N Saha
ZUSAMMENFASSUNG
The objective of this study was to design oral controlled release (CR) matrix tablets of Milnacipran using hydroxypropyl methylcellulose (HPMC) as the retardant polymer and to study the effect of various formulation factors such as polymer proportion, polymer viscosity, compression force and also the pH of dissolution medium on the in-vitro release of drug. Two viscosity grade of HPMC (15 K and 100 K) were used in the proportion of 50, 100, 150 and 200 mg per CR tablet. In-vitro release rate was characterized using various model dependent approaches and model independent dissolution parameters [T50% and T80% dissolution time, mean dissolution time (MDT), mean residence time (MRT), dissolution efficiency (DE)]. The statistical analysis was performed on all the model independent approaches using student t test and ANOVA. Results were found that as polymer concentration (50 mg to 200 mg) and viscosity (15 K to 100 K) increases, the MDT, MRT, T50% and T80% extended significantly. Drug release rate was found to be significantly different at different hardness. In-vivo human plasma concentration--time profile was predicted from in-vitro release data using convolution method. Predicted human pharmacokinetic parameters shows that the design CR formulation has capability to sustained the plasma drug level of milnacipran.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylzellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methylzellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methylzellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methylzellulose, viscosity: 15 cP
Sigma-Aldrich
Methylzellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methylzellulose, viscosity: 25 cP
Sigma-Aldrich
Methylzellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methylzellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methocel® A15 LV, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methocel® MC, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methylzellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methylzellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP