Merck
  • Lymphatic transport of puerarin occurs after oral administration of different lipid-based formulations to unconscious lymph duct-cannulated rats.
Lymphatic transport of puerarin occurs after oral administration of different lipid-based formulations to unconscious lymph duct-cannulated rats.
Pharmaceutical development and technology (2013-08-28)
An Zhou, Tao Lu, Lei Wang, Chuanhua Lu, Lina Wang, Maolin Wan, Hongfei Wu
ZUSAMMENFASSUNG
This study investigated the potential of targeted intestinal lymphatic transport of puerarin via a lipid formulation approach. Three formulations of PEG nanoemulsion, nanosuspension and an oil suspension containing puerarin were examined with the lymph-cannulated anaesthetized rat model. Plasma and lymph samples were analyzed by HPLC. Lymph triglyceride was measured using an enzymatic colorimetric technique. After 8 h, the total administered dose accumulated in the thoracic lymph duct was analyze. Nanoemulsion, nanosuspension and oil suspension was 0.065 ± 0.006%, 0.137 ± 0.018%, 0.021 ± 0.002% of the administered dose, respectively. In nanoemulsion, nanosuspension and oil suspension group, the systemic bioavailability of oral puerarin was 11%, 16% and 11% for lymph-cannulated rats, 41%, 67% and 18% for control rats. Absorption into the intestinal lymph should thus contribute to ∼30%, 51% and 7% of the systemic bioavailable puerarin. This data indicated that lipid-based nano drug formulation produced higher lymph concentrations of puerarin than oil suspension. The nanosuspension formulation may be considerable in terms of increased local concentrations in lymphoid tissue.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Puerarin, ≥98.0% (HPLC)
Supelco
Puerarin, analytical standard