Merck
Trehalose is not a universal solution for solid lipid nanoparticles freeze-drying.
Pharmaceutical development and technology (2013-10-09)
Slavomira Doktorovova, Ranjita Shegokar, Lisete Fernandes, Paula Martins-Lopes, Amélia M Silva, Rainer H Müller, Eliana B Souto
ZUSAMMENFASSUNG
To prepare stable and easy to handle formulation of solid lipid nanoparticles (SLNs) by freeze-drying with or without cryoprotectants, as appropriate. SLNs were freeze-dried without cryoprotectants or with cryoprotectants in quantities selected by freeze-thaw test (sucrose, glucose) or literature search (trehalose, maltose). Appearance, re-dispersability and size distribution of re-dispersed samples were evaluated. SLN could be freeze-dried using 10% sucrose, trehalose or maltose. Trehalose was effective in protecting one of presented formulations that was already very stable on its own; its efficiency in protecting other two formulations was limited. Our results are in line with various reports of successful freeze-drying of SLN, but considering the stability of original dispersions, no improvement was achieved. We confirmed that trehalose is among the most suitable cryoprotectant for SLN, however it did not improve shelf-life of the most stable formulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
Maltose -Lösung, BioReagent, ~20% in H2O, Molecular Biology
Supelco
Saccharose, analytical standard, for enzymatic assay kit SCA20
Saccharose, European Pharmacopoeia (EP) Reference Standard