Merck
  • Applied choline-omics: lessons from human metabolic studies for the integration of genomics research into nutrition practice.
Applied choline-omics: lessons from human metabolic studies for the integration of genomics research into nutrition practice.
Journal of the Academy of Nutrition and Dietetics (2014-02-18)
Allyson A West, Marie A Caudill
ZUSAMMENFASSUNG
Nutritional genomics, defined as the study of reciprocal interactions among nutrients, metabolic intermediates, and the genome, along with other closely related nutritional -omic fields (eg, epigenomics, transcriptomics, and metabolomics) have become vital areas of nutrition study and knowledge. Utilizing results from human metabolic research on the essential nutrient choline, this article illustrates how nutrigenetic, nutrigenomic, and inter-related -omic research has provided new insights into choline metabolism and its effect on physiologic processes. Findings from highlighted choline research are also discussed in the context of translation to clinical and public health nutrition applications. Overall, this article underscores the utility of -omic research methods in elucidating nutrient metabolism as well as the potential for nutritional -omic concepts and discoveries to be broadly applied in nutritional practice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholinchlorid, ≥98%
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortison -Lösung, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Sigma-Aldrich
Cholinchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Cholinchlorid, ≥99%
Supelco
Cortisol -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholinchlorid, BioUltra, ≥99.0% (AT)
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard
Supelco
Cholinchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cholinchlorid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Hydrocortison für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Hydrocortison, European Pharmacopoeia (EP) Reference Standard