Merck
Protein transport by the bacterial Tat pathway.
Biochimica et biophysica acta (2014-03-04)
Roshani Patel, Sarah M Smith, Colin Robinson
ZUSAMMENFASSUNG
The twin-arginine translocation (Tat) system accomplishes the remarkable feat of translocating large - even dimeric - proteins across tightly sealed energy-transducing membranes. All of the available evidence indicates that it is unique in terms of both structure and mechanism; however its very nature has hindered efforts to probe the core translocation events. At the heart of the problem is the fact that two large sub-complexes are believed to coalesce to form the active translocon, and 'capturing' this translocation event has been too difficult. Nevertheless, studies on the individual components have come a long way in recent years, and structural studies have reached the point where educated guesses can be made concerning the most interesting aspects of Tat. In this article we review these studies and the emerging ideas in this field. This article is part of a Special Issue entitled: Protein trafficking and secretion in bacteria. Guest Editors: Anastassios Economou and Ross Dalbey.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Arginin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginin, reagent grade, ≥98%
Sigma-Aldrich
L-Arginin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginin, 99%, FCC, FG
SAFC
L-Arginin
Supelco
L-Arginin, Pharmaceutical Secondary Standard; Certified Reference Material
Arginin, European Pharmacopoeia (EP) Reference Standard