Merck
  • Comparison of peptide-major histocompatibility complex tetramers and dextramers for the identification of antigen-specific T cells.
Comparison of peptide-major histocompatibility complex tetramers and dextramers for the identification of antigen-specific T cells.
Clinical and experimental immunology (2014-03-29)
G Dolton, A Lissina, A Skowera, K Ladell, K Tungatt, E Jones, D Kronenberg-Versteeg, H Akpovwa, J M Pentier, C J Holland, A J Godkin, D K Cole, M A Neller, J J Miles, D A Price, M Peakman, A K Sewell
ZUSAMMENFASSUNG
Fluorochrome-conjugated peptide-major histocompatibility complex (pMHC) multimers are widely used for flow cytometric visualization of antigen-specific T cells. The most common multimers, streptavidin-biotin-based 'tetramers', can be manufactured readily in the laboratory. Unfortunately, there are large differences between the threshold of T cell receptor (TCR) affinity required to capture pMHC tetramers from solution and that which is required for T cell activation. This disparity means that tetramers sometimes fail to stain antigen-specific T cells within a sample, an issue that is particularly problematic when staining tumour-specific, autoimmune or MHC class II-restricted T cells, which often display TCRs of low affinity for pMHC. Here, we compared optimized staining with tetramers and dextramers (dextran-based multimers), with the latter carrying greater numbers of both pMHC and fluorochrome per molecule. Most notably, we find that: (i) dextramers stain more brightly than tetramers; (ii) dextramers outperform tetramers when TCR-pMHC affinity is low; (iii) dextramers outperform tetramers with pMHC class II reagents where there is an absence of co-receptor stabilization; and (iv) dextramer sensitivity is enhanced further by specific protein kinase inhibition. Dextramers are compatible with current state-of-the-art flow cytometry platforms and will probably find particular utility in the fields of autoimmunity and cancer immunology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, 25,000
USP
Biotin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 150,000
Supelco
Dextran, analytical standard, 5,000
Supelco
Dextran aus Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 670,000
Supelco
Dextran, analytical standard, suitable for gel permeation chromatography (GPC), Set Mp 1,000-400,000
SAFC
BIOTIN
Supelco
Dextran, analytical standard, 670,000
Supelco
Dextran, analytical standard, 150,000
Supelco
Dextran, analytical standard, 1,000
Supelco
Dextran, analytical standard, 270,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran, analytical standard, 12,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran -Lösung aus Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, Mr ~60,000
Supelco
Dextran, analytical standard, 410,000