Merck
  • Spermidine/spermine N(1)-acetyltransferase activity associates with white blood cell count in myeloid leukemias.
Spermidine/spermine N(1)-acetyltransferase activity associates with white blood cell count in myeloid leukemias.
Experimental Hematology (2014)
Pirnes-Karhu S, Jantunen E, Mantymaa P, et al.