Merck
  • Ammonia and formaldehyde participate in the formation of 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) in addition to creati(ni)ne and phenylacetaldehyde.
Ammonia and formaldehyde participate in the formation of 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) in addition to creati(ni)ne and phenylacetaldehyde.
Food chemistry (2014-03-07)
Rosario Zamora, Esmeralda Alcón, Francisco J Hidalgo
ZUSAMMENFASSUNG
The formation of formaldehyde from phenylacetaldehyde and phenylalanine, and the contribution of both formaldehyde and ammonia to the production of PhIP from phenylacetaldehyde and creatinine were studied in an attempt to clarify the reaction pathways that produce PhIP. Formaldehyde was produced by thermal degradation of phenylacetaldehyde and, to a lesser extent, also by degradation of phenylalanine, phenylethylamine, styrene, and creatinine. When formaldehyde was added to a mixture of phenylacetaldehyde and creatinine, PhIP yield was multiplied by nineteen. When formaldehyde and ammonia were simultaneously present, PhIP yield was multiplied by fifty and the Ea of the reaction decreased by 61%. All these results point to formaldehyde and ammonia as the two additional reactants required for PhIP formation from both phenylacetaldehyde/creati(ni)ne and phenylalanine/creati(ni)ne mixtures. A general pathway for PhIP formation is proposed. This pathway is suggested to be the main route for PhIP formation in foods.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Acetaldehyd, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyd, natural, FG
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in ethanol
Sigma-Aldrich
Acetaldehyd, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
Ammoniak, anhydrous, ≥99.98%
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in dioxane
Supelco
Acetaldehyd, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyd, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Ammoniak -Lösung, 4 M in methanol
Sigma-Aldrich
Phenylacetaldehyd, ≥90%
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in THF
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in isopropanol
Sigma-Aldrich
Phenylacetaldehyd, ≥95%, FCC, FG
Sigma-Aldrich
Acetaldehyd -Lösung, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyd -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.
Sigma-Aldrich
Ammoniak, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Acetaldehyd -Lösung, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyd -Lösung, 5 M in THF
Sigma-Aldrich
Acetaldehyd -Lösung, 40 wt. % in isopropanol