Merck
  • Regulation of brain-type creatine kinase by AMP-activated protein kinase: interaction, phosphorylation and ER localization.
Regulation of brain-type creatine kinase by AMP-activated protein kinase: interaction, phosphorylation and ER localization.
Biochimica et biophysica acta (2014-04-15)
Sacnicte Ramírez Ríos, Frédéric Lamarche, Cécile Cottet-Rousselle, Anna Klaus, Roland Tuerk, Ramon Thali, Yolanda Auchli, René Brunisholz, Dietbert Neumann, Luc Barret, Malgorzata Tokarska-Schlattner, Uwe Schlattner
ZUSAMMENFASSUNG
AMP-activated protein kinase (AMPK) and cytosolic brain-type creatine kinase (BCK) cooperate under energy stress to compensate for loss of adenosine triphosphate (ATP) by either stimulating ATP-generating and inhibiting ATP-consuming pathways, or by direct ATP regeneration from phosphocreatine, respectively. Here we report on AMPK-dependent phosphorylation of BCK from different species identified by in vitro screening for AMPK substrates in mouse brain. Mass spectrometry, protein sequencing, and site-directed mutagenesis identified Ser6 as a relevant residue with one site phosphorylated per BCK dimer. Yeast two-hybrid analysis revealed interaction of active AMPK specifically with non-phosphorylated BCK. Pharmacological activation of AMPK mimicking energy stress led to BCK phosphorylation in astrocytes and fibroblasts, as evidenced with a highly specific phospho-Ser6 antibody. BCK phosphorylation at Ser6 did not affect its enzymatic activity, but led to the appearance of the phosphorylated enzyme at the endoplasmic reticulum (ER), close to the ER calcium pump, a location known for muscle-type cytosolic creatine kinase (CK) to support Ca²⁺-pumping.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Kreatinphosphokinase aus Kaninchenmuskel, Type I, salt-free, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
L-Serin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Creatine Phosphokinase from bovine heart, Type III, salt-free, lyophilized powder, ≥30 units/mg protein
Supelco
L-Serin, Pharmaceutical Secondary Standard; Certified Reference Material
Serin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Serin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland