Merck
  • Lysylated phospholipids stabilize models of bacterial lipid bilayers and protect against antimicrobial peptides.
Lysylated phospholipids stabilize models of bacterial lipid bilayers and protect against antimicrobial peptides.
Biochimica et biophysica acta (2014-05-02)
Elizabeth Cox, Austen Michalak, Sarah Pagentine, Pamela Seaton, Antje Pokorny
ZUSAMMENFASSUNG
Aminoacylated phosphatidylglycerols are common lipids in bacterial cytoplasmic membranes. Their presence in Staphylococcus aureus has been linked to increased resistance to a number of antibacterial agents, including antimicrobial peptides. Most commonly, the phosphatidylglycerol headgroup is esterified to lysine, which converts anionic phosphatidylglycerol into a cationic lipid with a considerably increased headgroup size. In the present work, we investigated the interactions of two well-studied antimicrobial peptides, cecropin A and mastoparan X, with lipid vesicles composed of 1-palmitoyl-2-oleoyl-phosphatidylcholine (POPC) and 1-palmitoyl-2-oleoyl-phosphatidylglycerol (POPG), containing varying fractions of an aminoacylated phosphatidylethanolamine, a stable analog of the corresponding phosphatidylglycerol-derivative. To differentiate between the effects of headgroup size and charge on peptide-lipid interactions, we synthesized two different derivatives. In one, the headgroup was modified by the addition of lysine, and in the other, by glutamine. The modification by glutamine results in a phospholipid with a headgroup size comparable to that of the lysylated version. However, whereas lysyl-phosphatidylethanolamine (Lys-PE) is cationic, glutaminyl-phosphatidylethanolamine (Gln-PE) is zwitterionic. We found that binding of mastoparan X and cecropin A was not significantly altered if the content of aminoacylated phosphatidylethanolamines did not exceed 20mol.%, which is the concentration found in bacterial membranes. However, a lysyl-phosphatidylethanolamine content of 20mol% significantly inhibits dye release from lipid vesicles, to a degree that depends on the peptide. In the case of mastoparan X, dye release is essentially abolished at 20mol.% lysyl-phosphatidylethanolamine, whereas cecropin A is less sensitive to the presence of lysyl-phosphatidylethanolamine. These observations are understood through the complex interplay between peptide binding and membrane stabilization as a function of the aminoacylated lipid content. This article is part of a Special Issue entitled: Interfacially Active Peptides and Proteins. Guest Editors: William C. Wimley and Kalina Hristova.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
L-Lysin, ≥98% (TLC)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysin, crystallized, ≥98.0% (NT)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium-hexafluorophosphat, ≥98.0% (T)
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
L-Lysin, analytical standard
Supelco
L-Lysin -monohydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)
Lysin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Lysin -acetat (Salz), ≥98% (HPLC)
Sigma-Aldrich
Cecropin A, ≥97% (HPLC), powder
Supelco
L-Lysin -monohydrochlorid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysinacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Lysin-acetat, European Pharmacopoeia (EP) Reference Standard