Merck
  • Uracil excision by endogenous SMUG1 glycosylase promotes efficient Ig class switching and impacts on A:T substitutions during somatic mutation.
Uracil excision by endogenous SMUG1 glycosylase promotes efficient Ig class switching and impacts on A:T substitutions during somatic mutation.
European journal of immunology (2014-04-29)
Felix A Dingler, Kristin Kemmerich, Michael S Neuberger, Cristina Rada
ZUSAMMENFASSUNG
Excision of uracil introduced into the immunoglobulin loci by AID is central to antibody diversification. While predominantly carried out by the UNG uracil-DNA glycosylase as reflected by deficiency in immunoglobulin class switching in Ung(-/-) mice, the deficiency is incomplete, as evidenced by the emergence of switched IgG in the serum of Ung(-/-) mice. Lack of switching in mice deficient in both UNG and MSH2 suggested that mismatch repair initiated a backup pathway. We now show that most of the residual class switching in Ung(-/-) mice depends upon the endogenous SMUG1 uracil-DNA glycosylase, with in vitro switching to IgG1 as well as serum IgG3, IgG2b, and IgA greatly diminished in Ung(-/-) Smug1(-/-) mice, and that Smug1 partially compensates for Ung deficiency over time. Nonetheless, using a highly MSH2-dependent mechanism, Ung(-/-) Smug1(-/-) mice can still produce detectable levels of switched isotypes, especially IgG1. While not affecting the pattern of base substitutions, SMUG1 deficiency in an Ung(-/-) background further reduces somatic hypermutation at A:T base pairs. Our data reveal an essential requirement for uracil excision in class switching and in facilitating noncanonical mismatch repair for the A:T phase of hypermutation presumably by creating nicks near the U:G lesion recognized by MSH2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ammoniumchlorid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Ammoniumchlorid, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammoniumchlorid, 99.998% trace metals basis
Supelco
Ammoniumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammoniumchlorid, 99.99% trace metals basis
Sigma-Aldrich
Ammoniumchlorid, BioUltra, ≥99.5% (AT)
Supelco
Ammonium-Ionenlösung für ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Ammoniumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium-14N-chlorid, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Fluorouracil Unreinheit C, European Pharmacopoeia (EP) Reference Standard