Merck
  • Does prolonged pituitary down-regulation with gonadotropin-releasing hormone agonist improve the live-birth rate in in vitro fertilization treatment?
Does prolonged pituitary down-regulation with gonadotropin-releasing hormone agonist improve the live-birth rate in in vitro fertilization treatment?
Fertility and sterility (2014-04-22)
Jianzhi Ren, Aiguo Sha, Dongmei Han, Ping Li, Jie Geng, Chaihui Ma
ZUSAMMENFASSUNG
To evaluate the effects of a prolonged duration of gonadotropin-releasing hormone agonist (GnRH-a) in pituitary down-regulation for controlled ovarian hyperstimulation (COH) on the live-birth rate in nonendometriotic women undergoing in vitro fertilization and embryo transfer (IVF-ET). Retrospective cohort study. University-affiliated hospital. Normogonadotropic women undergoing IVF. Three hundred seventy-eight patients receiving a prolonged pituitary down-regulation with GnRH-a before ovarian stimulation and 422 patients receiving a GnRH-a long protocol. Live-birth rate per fresh ET. In comparison with the long protocol, the prolonged down-regulation protocol required a higher total dose of gonadotropins. A lower serum luteinizing hormone (LH) level on the starting day of gonadotropin and the day of human chorionic gonadotropin (hCG) and a fewer number of oocytes and embryos were observed in the prolonged down-regulation protocol. However, the duration of stimulation and number of high-quality embryos were comparable between the two groups. A statistically significantly higher implantation rate (50.27% vs. 39.69%), clinical pregnancy rate (64.02% vs. 56.87%) and live-birth rate per fresh transfer cycle (55.56% vs. 45.73%) were observed in the prolonged protocol. Prolonged down-regulation in a GnRH-a protocol might increase the live-birth rates in normogonadotropic women.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Gonadorelin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
[D-Trp6]-LH-RH, ≥97% (HPLC), powder