Merck
Conserved residues that modulate protein trans-splicing of Npu DnaE split intein.
The Biochemical journal (2014-04-25)
Qin Wu, Zengqiang Gao, Yong Wei, Guolin Ma, Yuchuan Zheng, Yuhui Dong, Yangzhong Liu
ZUSAMMENFASSUNG
The first crystal trans-structure of a naturally occurring split intein has been determined for the Npu (Nostoc punctiforme PCC73102) DnaE split intein. Guided by this structure, the residues NArg50 and CSer35, well conserved in DnaE split inteins, are identified to be critical in the trans-splicing of Npu DnaE split intein. An in vitro splicing assay demonstrates that NArg50 and CSer35 play synergistic roles in modulating its intein activity. The C-terminal CAsn36 exhibits two orientations of its side chain and interacts with both NArg50 and CSer35 through hydrogen bonding. These interactions likely facilitate the cyclization of asparagine in the course of protein splicing. The mutation of either residue reduces intein activity, and correlates with the low activity of the Ssp (Cyanobacterium synechocystis sp. strain PCC6803) DnaE split intein. On the other hand, NArg50 also forms a hydrogen bond with the highly conserved F-block CAsp17, thus influencing the N-S acyl shift during N-terminal cleavage. Sequence alignments show that residues NArg50 and CSer35 are rather conserved in those split inteins that lack a penultimate histidine residue. The conserved non-catalytic residues of split inteins modulate the efficiency of protein trans-splicing by hydrogen-bond interactions with the catalytic residues at the splice junction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Asparaginsäure, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Arginin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginin, reagent grade, ≥98%
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Asparaginsäure, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginin, 99%, FCC, FG
SAFC
L-Arginin
Sigma-Aldrich
L-Asparaginsäure, BioXtra, ≥99% (HPLC)
Supelco
L-Arginin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Asparaginsäure, ≥99% (TLC)
Sigma-Aldrich
L-Serin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Asparaginsäure, BioUltra, ≥99.5% (T)
SAFC
L-Asparaginsäure
Sigma-Aldrich
L-Asparaginsäure Kaliumsalz, ≥98% (HPLC)
Arginin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Asparaginsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serin, Pharmaceutical Secondary Standard; Certified Reference Material
Asparaginsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Asparaginsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Serin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Serin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland