Merck
  • Tissue plasminogen activator promotes postischemic neutrophil recruitment via its proteolytic and nonproteolytic properties.
Tissue plasminogen activator promotes postischemic neutrophil recruitment via its proteolytic and nonproteolytic properties.
Arteriosclerosis, thrombosis, and vascular biology (2014-04-26)
Bernd Uhl, Gabriele Zuchtriegel, Daniel Puhr-Westerheide, Marc Praetner, Markus Rehberg, Matthias Fabritius, Maximilian Hessenauer, Martin Holzer, Andrej Khandoga, Robert Fürst, Stefan Zahler, Fritz Krombach, Christoph A Reichel
ZUSAMMENFASSUNG
Neutrophil infiltration of the postischemic tissue considerably contributes to organ dysfunction on ischemia/reperfusion injury. Beyond its established role in fibrinolysis, tissue-type plasminogen activator (tPA) has recently been implicated in nonfibrinolytic processes. The role of this serine protease in the recruitment process of neutrophils remains largely obscure. Using in vivo microscopy on the postischemic cremaster muscle, neutrophil recruitment and microvascular leakage, but not fibrinogen deposition at the vessel wall, were significantly diminished in tPA(-/-) mice. Using cell transfer techniques, leukocyte and nonleukocyte tPA were found to mediate ischemia/reperfusion-elicited neutrophil responses. Intrascrotal but not intra-arterial application of recombinant tPA induced a dose-dependent increase in the recruitment of neutrophils, which was significantly higher compared with stimulation with a tPA mutant lacking catalytic activity. Whereas tPA-dependent transmigration of neutrophils was selectively reduced on the inhibition of plasmin or gelatinases, neutrophil intravascular adherence was significantly diminished on the blockade of mast cell activation or lipid mediator synthesis. Moreover, stimulation with tPA caused a significant elevation in the leakage of fluorescein isothiocyanate dextran to the perivascular tissue, which was completely abolished on neutrophil depletion. In vitro, tPA-elicited macromolecular leakage of endothelial cell layers was abrogated on the inhibition of its proteolytic activity. Endogenously released tPA promotes neutrophil transmigration to reperfused tissue via proteolytic activation of plasmin and gelatinases. As a consequence, tPA on transmigrating neutrophils disrupts endothelial junctions allowing circulating tPA to extravasate to the perivascular tissue, which, in turn, amplifies neutrophil recruitment through the activation of mast cells and release of lipid mediators.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fibrinogen aus Rinderplasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
Fibrinogen aus Humanplasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
Fibrinogen aus Humanplasma, 35-65% protein (≥90% of protein is clottable).
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)