Merck
  • Assessment of rose bengal versus riboflavin photodynamic therapy for inhibition of fungal keratitis isolates.
Assessment of rose bengal versus riboflavin photodynamic therapy for inhibition of fungal keratitis isolates.
American journal of ophthalmology (2014-05-06)
Alejandro Arboleda, Darlene Miller, Florence Cabot, Mukesh Taneja, Mariela C Aguilar, Karam Alawa, Guillermo Amescua, Sonia H Yoo, Jean-Marie Parel
ZUSAMMENFASSUNG
To compare the in vitro effect of rose bengal and riboflavin as photosensitizing agents for photodynamic therapy (PDT) on fungal isolates that are common causes of fungal keratitis. Experimental study. Three isolates (Fusarium solani, Aspergillus fumigatus, Candida albicans) recovered from patients with confirmed fungal keratitis were used in the experiments. Isolates were grown on Sabouraud-Dextrose agar, swabbed, and prepared in suspension, and 1 mL aliquots were inoculated onto test plates in triplicate. Test plates were separated into 5 groups: Group 1, no treatment; Group 2, 0.1% rose bengal alone; Group 3, 518 nm irradiation alone; Group 4, riboflavin PDT (riboflavin + 375 nm irradiation); and Group 5, rose bengal PDT (rose bengal + 518 nm irradiation). Irradiation was performed over a circular area using either a green light-emitting diode (LED) array (peak wavelength: 518 nm) or an ultraviolet-A LED array (peak wavelength: 375 nm). Test plates were irradiated with an energy density of 5.4 J/cm(2). Later, plates were placed in a 30 C incubator and observed for growth. Rose bengal-mediated PDT successfully inhibited the growth of all 3 fungal isolates in the irradiated area. All other groups exhibited unrestricted growth throughout the plate. Rose bengal-mediated PDT successfully inhibited the growth of 3 types of fungi. No other experimental groups, including riboflavin-mediated PDT, had any inhibitory effect on the isolates. The results might be useful for the treatment of patients suffering from corneal infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Supelco
(−)-Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Riboflavin 5′-phosphate sodium salt hydrate, meets USP testing specifications
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Riboflavin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard