Merck
Induction of insulin secretion by apolipoprotein M, a carrier for sphingosine 1-phosphate.
Biochimica et biophysica acta (2014-05-13)
Makoto Kurano, Masumi Hara, Koichi Tsuneyama, Hideyuki Sakoda, Tomo Shimizu, Kazuhisa Tsukamoto, Hitoshi Ikeda, Yutaka Yatomi
ZUSAMMENFASSUNG
High-density lipoprotein (HDL) has been proposed to enhance β-cell functions. Clinical studies have suggested that apolipoprotein M (apoM), which rides mainly on HDL, is involved in diabetes; however, the underlying mechanism has not yet been elucidated. Recently, apoM was shown to be a carrier for sphingosine 1-phosphate (S1P), a bioactive lipid mediator. In the present study, we investigated the modulation of insulin secretion by apoM through the action of S1P. We overexpressed apoM in the livers of C57BL6 mice using adenovirus gene transfer and found that the blood glucose levels under ad libitum feeding conditions were lower in the apoM-overexpressing mice. While an insulin tolerance test revealed that insulin sensitivity was not significantly affected, a glucose tolerance test revealed that apoM-overexpressing mice had a better glucose tolerance because of enhanced insulin secretion, a phenomenon that was reversed by treatment with VPC 23019, an antagonist against S1P1 and S1P3 receptor. In vitro experiments with MIN6 cells also revealed that apoM-containing lipoproteins enhanced insulin secretion, which was again inhibited by VPC 23019. ApoM retarded the degradation of S1P, and an increase in Pdx1 expression, the attenuation of endoreticulum stress, and the phosphorylation of Akt, AmpK, and Erk were observed as possible underlying mechanisms for the effect of S1P, maintained at a high concentration by apoM, on the increase in insulin secretion. ApoM augmented insulin secretion by maintaining the S1P concentration under both in vivo and in vitro conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sphingosin-1-phosphat, ≥95%, powder
Sigma-Aldrich
Sphingosin-1-phosphat, ≥98.0% (TLC)
Sigma-Aldrich
D-Sphingosin, synthetic