Merck
  • Functional effects of alcohol withdrawal syndrome on peripheral sympathetic neurotransmission in vas deferens of adult rats.
Functional effects of alcohol withdrawal syndrome on peripheral sympathetic neurotransmission in vas deferens of adult rats.
Life sciences (2014-05-17)
Guilherme Henrique Souza Bomfim, Luciana Ferreira Verde, Roberto Frussa-Filho, Aron Jurkiewicz, Neide Hyppolito Jurkiewicz
ZUSAMMENFASSUNG
Alcohol withdrawal syndrome (AWS) is characterized by a set of physiological modifications triggered by abrupt withdrawal and/or decreasing consumption of ethanol (EtOH), which may manifest 16-48 h after ceasing consumption. The relationship between the effects of AWS and central and peripheral sympathetic neurotransmission is unknown. This study investigates the possible mechanisms on the sympathetic system during periods of AWS. Male Wistar rats were treated with EtOH (6-10 g/kg/day/v.o. 5 days). Subsequently, 1h, 24h, 48 h and 120 h after administration of the last dose of EtOH, the animals were sacrificed, and their vas deferens (VD) were removed to perform the following evaluations: (a) concentration-effect curves of sympathetic agonist; (b) activity of α2-adrenoreceptor; (c) function of voltage-dependent calcium channels (Cav); and (d) release of endogenous catecholamines measured in real time coupled to HPLC. The results showed that the maximum effects of contraction were increased by agonists tested in at 24h and 48 h EtOH withdrawal. The inhibitory affinity (pIC50) of guanfacine was decreased 24h EtOH withdrawal. The function of Cav was also decreased as pIC50 values dropped 24h and 48 h EtOH withdrawal. The release of catecholamines increased 48 h after EtOH withdrawal. It is suggested that AWS triggers hyperactivity in peripheral sympathetic neurotransmission. The mechanisms underlying hyperactivity are possibly explained by a failure of autoregulation from catecholamines released by α2-adrenoreceptors and/or an increase of Cav function, which may be potential targets to attenuate the symptoms of AWS at the peripheral level.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 Ampullen/Kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 Ampullen/Kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol -Lösung, 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 Ampullen/Kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Guanfacin -hydrochlorid, ≥98% (HPLC)
Supelco
Ethanol -Lösung, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®