Merck
Preventive treatment for tuberculosis in people with HIV.
Lancet (London, England) (2014-05-20)
Soumya Swaminathan
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Isoniazid, European Pharmacopoeia (EP) Reference Standard