Merck
  • Mouse model for deficiency of methionine synthase reductase exhibits short-term memory impairment and disturbances in brain choline metabolism.
Mouse model for deficiency of methionine synthase reductase exhibits short-term memory impairment and disturbances in brain choline metabolism.
The Biochemical Journal (2014)
Jadavji NM, Bahous RH, Deng L, et al.