- Complete genome sequencing and comparative analysis of the linezolid-resistant Enterococcus faecalis strain DENG1.
Complete genome sequencing and comparative analysis of the linezolid-resistant Enterococcus faecalis strain DENG1.
Archives of microbiology (2014-05-08)
Zhijian Yu, Zhong Chen, Hang Cheng, Jinxin Zheng, Duoyun Li, Xiangbin Deng, Weiguang Pan, Weizhi Yang, Qiwen Deng
PMID24800692
ZUSAMMENFASSUNG
Genome level analysis of bacterial strains provides information on genetic composition and resistance mechanisms to clinically relevant antibiotics. To date, whole genome characterization of linezolid-resistant Enterococcus faecalis isolated in the clinic is lacking. In this study, we report the entire genome sequence, genomic characteristics and virulence factors of a pathogenic E. faecalis strain, DENG1. Our results showed considerable differences in genomic characteristics and virulence factors compared with other E. faecalis strains (V583 and OG1RF). The genome of this LZD-resistant E. faecalis strain can be used as a reference to study the mechanism of LZD resistance and the phylogenetic relationship of E. faecalis strains worldwide.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)