Merck
  • Oxidative modification of fibrinogen is associated with altered function and structure in the subacute phase of myocardial infarction.
Oxidative modification of fibrinogen is associated with altered function and structure in the subacute phase of myocardial infarction.
Arteriosclerosis, thrombosis, and vascular biology (2014-05-03)
Matteo Becatti, Rossella Marcucci, Giulia Bruschi, Niccolò Taddei, Daniele Bani, Anna Maria Gori, Betti Giusti, Gian Franco Gensini, Rosanna Abbate, Claudia Fiorillo
ZUSAMMENFASSUNG
Among plasma proteins, fibrinogen represents a major target of oxidative modifications. In patients with post-acute myocardial infarction (6 months after the acute event), fibrinogen oxidation-induced carbonyls and fibrinogen function were estimated using in vitro and ex vivo approaches. Fibrinogen structural features and clot architecture were also explored. In 39 patients with post-acute myocardial infarction and 28 age-, sex-, and risk factor-matched controls, oxidative stress markers (in plasma and in purified fibrinogen fractions), thrombin-catalyzed fibrin polymerization, and plasmin-induced fibrin lysis were estimated. Circular dichroism spectra of purified fibrinogen extracts, electron microscopy, and differential interference contrast microscopy analyses of fibrin clots were also performed. Marked signs of oxidative stress in plasma (P<0.01 versus controls) and, correspondingly, an increased extent of fibrinogen carbonylation (3.5-fold over control values; P<0.01 versus controls) were observed in patients. Furthermore, fibrinogen fractions purified from patients exhibited significantly reduced clotting ability and decreased susceptibility to plasmin-induced lysis (P<0.01 versus controls). Alterations in fibrinogen secondary structure, as suggested by circular dichroism spectroscopy, and in fibrin clot architecture, as analyzed by electron and differential interference contrast microscopy, were also identified. Here, we report for the first time that patients with post-acute myocardial infarction present with an overall imbalance in redox status and marked fibrinogen carbonylation associated with altered fibrinogen function, thus suggesting a role for carbonylation as a direct mechanism of fibrinogen function. The observed features occur along with modifications in protein structure and in clot architecture.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fibrinogen aus Rinderplasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
Fibrinogen aus Humanplasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
Fibrinogen aus Humanplasma, 35-65% protein (≥90% of protein is clottable).
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)