Merck
Chitosan and silver nanoparticles: promising anti-toxoplasma agents.
Experimental parasitology (2014-05-24)
M R Gaafar, R F Mady, R G Diab, Th I Shalaby
ZUSAMMENFASSUNG
Toxoplasmosis is a worldwide infection caused by obligate intracellular protozoan parasite which is Toxoplasma gondii. Chitosan and silver nanoparticles were synthesized to be evaluated singly or combined for their anti-toxoplasma effects as prophylaxis and as treatment in the experimental animals. Results were assessed through studying the parasite density and the ultrastructural parasite changes, and estimation of serum gamma interferon. Weight of tissue silver was assessed in different organs. Results showed that silver nanoparticles used singly or combined with chitosan have promising anti-toxoplasma potentials. The animals that received these compounds showed statistically significant decrease in the mean number of the parasite count in the liver and the spleen, when compared to the corresponding control group. Light microscopic examination of the peritoneal exudates of animals receiving these compounds showed stoppage of movement and deformity in shape of the tachyzoites, whereas, by scanning electron microscope, the organisms were mutilated. Moreover, gamma interferon was increased in the serum of animals receiving these compounds. All values of silver detected in different tissues were within the safe range. Thus, these nanoparticles proved their effectiveness against the experimental Toxoplasma infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Silber, conductive paste
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Silber, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silber, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Chitosan aus Garnelenschalen, low-viscous
Sigma-Aldrich
Silber, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silber, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silber-Nanodrähte, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silber, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silber-Nanodrähte, diam. × L 200  nm × 10 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silber, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silber, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silber, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber-Nanodrähte, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silber, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silber, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silber, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silber, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silber, powder, <250 μm, 99.99% trace metals basis
Silber, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95+%
Sigma-Aldrich
Silber, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, foil, thickness 0.025 mm, 99.9% trace metals basis
Silber, foil, 2m coil, thickness 0.025mm, annealed, 99.95+%