Merck
  • Cadmium supplement triggers endoplasmic reticulum stress response and cytotoxicity in primary chicken hepatocytes.
Cadmium supplement triggers endoplasmic reticulum stress response and cytotoxicity in primary chicken hepatocytes.
Ecotoxicology and environmental safety (2014-05-20)
Cheng-Cheng Shao, Nan Li, Zi-Wei Zhang, Jian Su, Shu Li, Jin-Long Li, Shi-Wen Xu
ZUSAMMENFASSUNG
Cadmium (Cd), a potent hepatotoxin, has been reported to induce endoplasmic reticulum (ER) stress in various cell types. However, whether such effect exists in bird is still unclear. To delineate the effects of Cd exposure on ER stress response, we examined the expression of 78-kDa glucose-regulated protein (GRP78) and alteration in calcium homeostasis in primary chicken hepatocytes treated with 2-22 µM Cd for 24 h. A significant decrease of cell viability was observed in chicken hepatocytes following Cd administration. In cells treated with Cd, GRP78 protein levels increased in a dose-dependent manner. In addition, GRP78 and GRP94mRNA levels were elevated in response to Cd exposure. The increase of the intracellular Ca(2+) concentration in chicken hepatocytes was found during Cd exposure. Cd significantly decreased the CaM mRNA levels in hepatocytes. These results show that Cd regulates the expression of GRP78 and calcium homeostasis in chicken hepatocytes, suggesting that ER stress induced by Cd plays an important role in the mechanisms of Cd cytotoxicity to the bird hepatocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, rod, 100mm, diameter 40mm, 99.9%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, rod, 200mm, diameter 10.0mm, 99.999%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, foil, 30x50mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 50x50mm, thickness 2.0mm, as rolled, 99.99%
Cadmium, foil, 4mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 8mm disks, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, wire reel, 0.1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%