Merck
  • DDI-CPI, a server that predicts drug-drug interactions through implementing the chemical-protein interactome.
DDI-CPI, a server that predicts drug-drug interactions through implementing the chemical-protein interactome.
Nucleic acids research (2014-05-31)
Heng Luo, Ping Zhang, Hui Huang, Jialiang Huang, Emily Kao, Leming Shi, Lin He, Lun Yang
ZUSAMMENFASSUNG
Drug-drug interactions (DDIs) may cause serious side-effects that draw great attention from both academia and industry. Since some DDIs are mediated by unexpected drug-human protein interactions, it is reasonable to analyze the chemical-protein interactome (CPI) profiles of the drugs to predict their DDIs. Here we introduce the DDI-CPI server, which can make real-time DDI predictions based only on molecular structure. When the user submits a molecule, the server will dock user's molecule across 611 human proteins, generating a CPI profile that can be used as a feature vector for the pre-constructed prediction model. It can suggest potential DDIs between the user's molecule and our library of 2515 drug molecules. In cross-validation and independent validation, the server achieved an AUC greater than 0.85. Additionally, by investigating the CPI profiles of predicted DDI, users can explore the PK/PD proteins that might be involved in a particular DDI. A 3D visualization of the drug-protein interaction will be provided as well. The DDI-CPI is freely accessible at http://cpi.bio-x.cn/ddi/.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sertralin -hydrochlorid, ≥98% (HPLC)
USP
Sertralin -hydrochlorid, United States Pharmacopeia (USP) Reference Standard
Supelco
Sertraline hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sertralin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Sertralin für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Sertralin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard