Merck
  • Fluctuation of Dof1/Dof2 expression ratio under the influence of varying nitrogen and light conditions: involvement in differential regulation of nitrogen metabolism in two genotypes of finger millet (Eleusine coracana L.).
Fluctuation of Dof1/Dof2 expression ratio under the influence of varying nitrogen and light conditions: involvement in differential regulation of nitrogen metabolism in two genotypes of finger millet (Eleusine coracana L.).
Gene (2014-05-31)
Supriya Gupta, Sanjay Mohan Gupta, Alok Kumar Gupta, Vikram Singh Gaur, Anil Kumar
ZUSAMMENFASSUNG
In order to gain insights into the mechanism of high nitrogen use efficiency (NUE) of finger millet (FM) the role of Dof2 transcription factor (TF), which is a repressor of genes involved in C/N metabolism was investigated. The partial cDNA fragment of EcDof2 (912-bp; GenBank acc. no. KF261117) was isolated and characterized from finger millet (FM) that showed 63% and 58% homology with Dof2 of Zea mays at nucleotide and protein level, respectively. Its expression studies were carried out along with the activator EcDof1 in two genotypes (GE3885, high protein genotype (HPG); GE1437, low protein genotype (LPG)) of FM differing in grain protein contents (13.8% and 6.2%) showed that EcDof2 is expressed in both shoot and root tissues with significantly (p≤0.05) higher expression in the roots. The diurnal expression of both EcDof1 and EcDof2 in shoots was differential having different time of peak expression indicating a differential response to diurnal condition. Under continuous dark conditions, expression of EcDof1 and EcDof2 oscillated in both the genotypes whereas on illumination, the fold expression of EcDof1 was higher as compared to EcDof2. Under increasing nitrate concentration, EcDof2 expression increases in roots and shoots of LPG while it remains unchanged in HPG. However, the EcDof1 expression was found to increase in both genotypes. Further, time kinetics studies under single nitrate concentration revealed that EcDof2 was repressed in the roots of both genotypes whereas EcDof1 oscillated with time. The EcDof1/EcDof2 ratio measured showed differential response under different light and nitrogen conditions. It was higher in the roots of HPG indicating higher activation of genes involved in N uptake and assimilation resulting in high grain protein accumulation. The results indicate that both light and nitrogen concentration influence Dof1 and Dof2 expression and suggests a complex pattern of regulation of genes influenced by these plant specific TFs. In nutshell, the Dof1/Dof2 ratio can serve as an index for measuring the N responsiveness and NUE of crops and can be further validated by Dof2 knock down approach.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Stickstoff, ≥99.998%