Merck
  • Comparison of synovial fluid, urine, and serum ion levels in metal-on-metal total hip arthroplasty at a minimum follow-up of 18 years.
Comparison of synovial fluid, urine, and serum ion levels in metal-on-metal total hip arthroplasty at a minimum follow-up of 18 years.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2014-05-21)
Richard Lass, Alexander Grübl, Alexander Kolb, David Stelzeneder, Alexander Pilger, Bernd Kubista, Alexander Giurea, Reinhard Windhager
ZUSAMMENFASSUNG
Diagnosis of adverse reactions to metal debris in metal-on-metal hip arthroplasty is a multifactorial process. Systemic ion levels are just one factor in the evaluation and should not be relied upon solely to determine the need for revision surgery. Furthermore, the correlation between cobalt or chromium serum, urine, or synovial fluid levels and adverse local tissue reactions is still incompletely understood. The hypothesis was that elevated serum and urine metal-ion concentrations are associated with elevated local metal-ion concentrations in primary total hip arthroplasties (THA) and with failure of metal-on-metal articulations in the long-term. In our present study, we evaluated these concentrations in 105 cementless THA with metal-on-metal articulating surfaces with small head diameter at a minimum of 18 years postoperatively. Spearman correlation showed a high correlation between the joint fluid aspirate concentration of cobalt and chromium with the serum cobalt (r = 0.81) and chromium level (r = 0.77) in patients with the THA as the only source of metal-ions. In these patients serum metal-ion analysis is a valuable method for screening. In patients with more than one source of metal or renal insufficiency additional investigations, like joint aspirations are an important tool for evaluation of wear and adverse tissue reactions in metal-on-metal THA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Chrom, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Kobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Chrom, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chrom, chips, 99.995% trace metals basis
Sigma-Aldrich
Kobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Kobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Kobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Kobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Chrom, chips, thickness ~2 mm, 99.5%
Chrom, rod, 25mm, diameter 2.0mm, 99.7+%
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, ≥99.99%
Chrom, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Sigma-Aldrich
Kobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Chrom, rod, 50mm, diameter 2.0mm, 99.7+%
Kobalt, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Chrom, rod, 10mm, diameter 2.0mm, 99.7+%
Kobalt, foil, 4mm disks, thickness 0.05mm, annealed, 99.9%
Kobalt, foil, 8mm disks, thickness 0.003mm, 99.9%
Chrom, bar, 100mm x 2mm x 2mm, 99.7+%
Kobalt, foil, 6mm disks, thickness 0.01mm, 99.9%
Kobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Kobalt, foil, 8mm disks, thickness 0.01mm, 99.9%
Kobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Kobalt, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Kobalt, foil, light tested, 50x50mm, thickness 0.0125mm, 99.9%
Kobalt, wire reel, 0.5m, diameter 0.05mm, hard, 99.9%
Kobalt, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%