Merck
Tet oxidizes thymine to 5-hydroxymethyluracil in mouse embryonic stem cell DNA.
Nature chemical biology (2014-05-20)
Toni Pfaffeneder, Fabio Spada, Mirko Wagner, Caterina Brandmayr, Silvia K Laube, David Eisen, Matthias Truss, Jessica Steinbacher, Benjamin Hackner, Olga Kotljarova, David Schuermann, Stylianos Michalakis, Olesea Kosmatchev, Stefan Schiesser, Barbara Steigenberger, Nada Raddaoui, Gengo Kashiwazaki, Udo Müller, Cornelia G Spruijt, Michiel Vermeulen, Heinrich Leonhardt, Primo Schär, Markus Müller, Thomas Carell
ZUSAMMENFASSUNG
Ten eleven translocation (Tet) enzymes oxidize the epigenetically important DNA base 5-methylcytosine (mC) stepwise to 5-hydroxymethylcytosine (hmC), 5-formylcytosine and 5-carboxycytosine. It is currently unknown whether Tet-induced oxidation is limited to cytosine-derived nucleobases or whether other nucleobases are oxidized as well. We synthesized isotopologs of all major oxidized pyrimidine and purine bases and performed quantitative MS to show that Tet-induced oxidation is not limited to mC but that thymine is also a substrate that gives 5-hydroxymethyluracil (hmU) in mouse embryonic stem cells (mESCs). Using MS-based isotope tracing, we show that deamination of hmC does not contribute to the steady-state levels of hmU in mESCs. Protein pull-down experiments in combination with peptide tracing identifies hmU as a base that influences binding of chromatin remodeling proteins and transcription factors, suggesting that hmU has a specific function in stem cells besides triggering DNA repair.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cytosin, ≥99%
Sigma-Aldrich
Thymin, ≥99%
Sigma-Aldrich
Thymin, BioReagent, suitable for cell culture
Supelco
Cytosin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-(Hydroxymethyl)uracil, 97%
Supelco
Thymin (Zidovudin-verwandte Verbindung C), Pharmaceutical Secondary Standard; Certified Reference Material
Gemcitabin Unreinheit A, European Pharmacopoeia (EP) Reference Standard