Merck
  • X-ray structure of a novel L-ribose isomerase acting on a non-natural sugar L-ribose as its ideal substrate.
X-ray structure of a novel L-ribose isomerase acting on a non-natural sugar L-ribose as its ideal substrate.
The FEBS journal (2014-05-23)
Hiromi Yoshida, Akihide Yoshihara, Misa Teraoka, Yuji Terami, Goro Takata, Ken Izumori, Shigehiro Kamitori
ZUSAMMENFASSUNG
l-Ribose, a pentose, is not known to exist in nature. Although organisms typically do not have a metabolic pathway that uses l-ribose as a carbon source, prokaryotes use various sugars as carbon sources for survival. Acinetobacter sp. DL-28 has been shown to express the novel enzyme, l-ribose isomerase (AcL-RbI), which catalyzes reversible isomerization between l-ribose and l-ribulose. AcL-RbI showed the highest activity to l-ribose, followed by d-lyxose with 47% activity, and had no significant amino acid sequence similarity to structure-known proteins, except for weak homology with the d-lyxose isomerases from Escherichia coli O157 : H7 (18%) and Bacillus subtilis strain (19%). Thus, AcL-RbI is expected to have the unique three-dimensional structure to recognize l-ribose as its ideal substrate. The X-ray structures of AcL-RbI in complexes with substrates were determined. AcL-RbI had a cupin-type β-barrel structure, and the catalytic site was found between two large β-sheets with a bound metal ion. The catalytic site structures clearly showed that AcL-RbI adopted a cis-enediol intermediate mechanism for the isomerization reaction using two glutamate residues (Glu113 and Glu204) as acid/base catalysts. In its crystal form, AcL-RbI formed a unique homotetramer with many substrate sub-binding sites, which likely facilitated capture of the substrate. The atomic coordinates and structure factors of AcL-RbI/l-ribose, AcL-RbI/l-ribulose, AcL-RbI/ribitol, E204Q/l-ribose and E204Q/l-ribulose have been deposited in the Protein Data Bank under accession codes, 4Q0P, 4Q0Q, 4Q0S, 4Q0U and 4Q0V.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutaminsäure, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutaminsäure, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
L-Glutaminsäure, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
D-Glutaminsäure, ≥99% (TLC)
Supelco
L-Glutaminsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
D-(−)-Ribose, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutaminsäure, FCC
Sigma-Aldrich
L-Glutaminsäure -hydrochlorid, ≥99% (HPLC)
Glutaminsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ribulose, ≥90% (HPLC)
Supelco
L-Glutaminsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland