Merck
  • Zinc enhances the phototoxic effect of blue light against malodour-producing bacteria in an experimental oral biofilm.
Zinc enhances the phototoxic effect of blue light against malodour-producing bacteria in an experimental oral biofilm.
Journal of medical microbiology (2014-06-11)
Nir Sterer, Uziel Jeffet, Aurel Dadoun, Ronit Bar-Ness Greenstein, David Kohavi
ZUSAMMENFASSUNG
Oral malodour is thought to be caused mainly by the production of volatile sulfide compounds (VSCs) by anaerobic Gram-negative oral bacteria. Previous studies have shown that these bacteria are susceptible to blue light (400-500 nm wavelength). In the present study, we tested the effect of blue light in the presence of zinc, erythrosine B or both on malodour production in an experimental oral biofilm. Biofilms were exposed to a plasma-arc light source for 30, 60 and 120 s (equal to energy fluxes of 41, 82 and 164 J cm(-2), respectively) with or without the addition of zinc acetate, erythrosine B or both. After the light exposure, biofilm samples were examined for malodour production (by an odour judge) and VSC production (with a Halimeter), and VSC-producing bacteria were quantified using a microscopy-based sulfide assay (MSA) and in situ confocal laser scanning microscopy (CLSM). Results showed that exposing experimental oral biofilm to both blue light and zinc reduced malodour production, which coincided with a reduction in VSC-producing bacteria in the biofilm. These results suggest that zinc enhances the phototoxicity of blue light against malodour-producing bacteria.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zink, dust, <10 μm, ≥98%
Sigma-Aldrich
Zink, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zink, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zink, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zink, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zink, purum, powder
Sigma-Aldrich
Zink, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zink, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Zink, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95+%
Sigma-Aldrich
Zink, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zink, mossy, ≥99%
Sigma-Aldrich
Zink Zubereitung, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zink, pieces, 2-14 mesh, 99.9% trace metals basis
Zink, wire reel, 5m, diameter 1.0mm, 99.99+%
Sigma-Aldrich
Zink, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zink, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zink, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zink, shot, 5 mm, 99.999% trace metals basis
Zink, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95+%
Sigma-Aldrich
Zink, foil, thickness 1.0 mm, 99.99% trace metals basis
Zink, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95+%
Zink, foil, 1m coil, thickness 0.025mm, as rolled, 99.95+%
Zink, foil, 5m coil, thickness 0.125mm, as rolled, 99.95+%
Zink, wire reel, 1m, diameter 1.0mm, 99.99+%
Zink, wire reel, 0.5m, diameter 1.0mm, 99.99+%
Zink, foil, 0.5m coil, thickness 0.05mm, as rolled, 99.95+%
Zink, foil, 100x100mm, thickness 0.15mm, as rolled, 99.95+%
Zink, foil, 300x300mm, thickness 2.0mm, as rolled, 99.95+%
Zink, wire reel, 25m, diameter 0.125mm, as drawn, 99%
Zink, wire reel, 10m, diameter 0.25mm, as drawn, 99.99+%