Merck
  • Chemokine (C-X-C motif) receptor 4 blockade by AMD3100 inhibits experimental abdominal aortic aneurysm expansion through anti-inflammatory effects.
Chemokine (C-X-C motif) receptor 4 blockade by AMD3100 inhibits experimental abdominal aortic aneurysm expansion through anti-inflammatory effects.
Arteriosclerosis, thrombosis, and vascular biology (2014-05-31)
Stéphanie Michineau, Grégory Franck, Orianne Wagner-Ballon, Jianping Dai, Eric Allaire, Marianne Gervais
ZUSAMMENFASSUNG
Inflammation plays a critical role in the development of abdominal aortic aneurysms (AAAs). Because stromal cell-derived factor 1 (SDF-1) is known for its ability to attract inflammatory cells, we investigated whether SDF-1/chemokine (C-X-C motif) receptor 4 (CXCR4) axis is expressed in aneurysmal aortic wall and plays a role in AAA physiopathology and asked whether its blockade modulates AAA formation and expansion. Quantitative real-time polymerase chain reaction analysis showed that SDF-1α and CXCR4 mRNA levels are increased in both human and CaCl2-induced mouse AAA wall and are positively correlated to the aortic diameter in mice. ELISA quantification and immunostaining demonstrated that, in mice, aortic SDF-1α is rapidly induced during AAA formation, first by apoptotic vascular smooth muscle cells in the injured media and then by adventitial macrophages once AAA is fully established. Using green fluorescent protein-positive (GFP(+/-)) bone marrow transplantation experiments, we demonstrated that aortic SDF-1 overexpression is implicated in the recruitment of bone marrow-derived macrophages within the AAA wall. Furthermore, in mice, blockade of CXCR4 by AMD3100 decreases the infiltration of adventitial macrophages, inhibits AAA formation, and prevents aortic wall destruction. AMD3100 reduces the mRNA levels of MMP-12 and MMP-14 as well as that of inflammatory effectors MCP-1, MIP-1β, MIP-2α, RANTES, IL-1β, IL-6, TNF-α, and E-selectin. Finally, AMD3100 stabilizes the diameter of formed, expanding AAAs in 2 experimental models. SDF-1/CXCR4 axis is upregulated in human and mouse AAAs. Blockade of CXCR4 with AMD3100 suppresses AAA formation and progression in two rodent models. Blockade of SDF-1/CXCR4 axis may represent a new strategy to limit progression of small human AAAs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calciumchlorid Dihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Calciumchlorid Dihydrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calciumchlorid Dihydrat, Molecular Biology, ≥99.0%
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calciumchlorid Dihydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Calciumchlorid Dihydrat, BioUltra, Molecular Biology, ≥99.5% (KT)
Sigma-Aldrich
Calciumchlorid Dihydrat, tested according to Ph. Eur.
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calciumchlorid Dihydrat, BioXtra, ≥99.0%
Sigma-Aldrich
AMD3100 -octahydrochlorid Hydrat, ≥97% (NMR), solid
Supelco
Calcium Standard für AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Supelco
Calcium Ionenlösung für ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis