Merck
  • A microplate assay for the coupled transglycosylase-transpeptidase activity of the penicillin binding proteins; a vancomycin-neutralizing tripeptide combination prevents penicillin inhibition of peptidoglycan synthesis.
A microplate assay for the coupled transglycosylase-transpeptidase activity of the penicillin binding proteins; a vancomycin-neutralizing tripeptide combination prevents penicillin inhibition of peptidoglycan synthesis.
Biochemical and biophysical research communications (2014-06-20)
Vidya P Kumar, Chandrakala Basavannacharya, Sunita M de Sousa
ZUSAMMENFASSUNG
A microplate, scintillation proximity assay to measure the coupled transglycosylase-transpeptidase activity of the penicillin binding proteins in Escherichia coli membranes was developed. Membranes were incubated with the two peptidoglycan sugar precursors UDP-N-acetyl muramylpentapeptide (UDP-MurNAc(pp)) and UDP-[(3)H]N-acetylglucosamine in the presence of 40 μM vancomycin to allow in situ accumulation of lipid II. In a second step, vancomycin inhibition was relieved by addition of a tripeptide (Lys-D-ala-D-ala) or UDP-MurNAc(pp), resulting in conversion of lipid II to cross-linked peptidoglycan. Inhibitors of the transglycosylase or transpeptidase were added at step 2. Moenomycin, a transglycosylase inhibitor, had an IC50 of 8 nM. Vancomycin and nisin also inhibited the assay. Surprisingly, the transpeptidase inhibitors penicillin and ampicillin showed no inhibition. In a pathway assay of peptidoglycan synthesis, starting from the UDP linked sugar precursors, inhibition by penicillin was reversed by a 'neutral' combination of vancomycin plus tripeptide, suggesting an interaction thus far unreported.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vancomycin -hydrochlorid aus Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Vancomycin -hydrochlorid aus Streptomyces orientalis, ≥85% (Vancomycin B)
Sigma-Aldrich
Vancomycin -hydrochlorid aus Streptomyces orientalis, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Vancomycin -hydrochlorid aus Streptomyces orientalis, meets USP testing specifications
Millipore
Vancomycin-Supplement, suitable for microbiology
Vancomycin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard