Merck
  • Efficient generation and increased reactivity in cationic gold via Brønsted acid or Lewis acid assisted activation of an imidogold precatalyst.
Efficient generation and increased reactivity in cationic gold via Brønsted acid or Lewis acid assisted activation of an imidogold precatalyst.
Organic letters (2014-06-24)
Junbin Han, Naoto Shimizu, Zhichao Lu, Hideki Amii, Gerald B Hammond, Bo Xu
ZUSAMMENFASSUNG
Brønsted or Lewis acid assisted activation of an imidogold precatalyst (L-Au-Pht, Pht = phthalimide) offers a superior way to generate cationic gold compared with the commonly used silver-based system. It is also broadly applicable for most common gold-catalyzed reactions. For reactions that require milder conditions, milder acids can be used for optimized efficiency.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Silber, conductive paste
Sigma-Aldrich
Silber, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silber, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silber, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silber, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silber, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Silber-Nanodrähte, diam. × L 200  nm × 10 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silber, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Silber, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Goldbeschichteter Glasobjektträger, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Silber, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silber, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silber, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silber, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Silber, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silber, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Goldbeschichteter Glasobjektträger, layer thickness 1000 Å, 99.999% (Au)
Silber, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95+%
Gold, rod, 50mm, diameter 3.0mm, as drawn, 99.95%
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis