Merck
  • ATP-binding mode including a carbamoylated lysine and two Mg(2+) ions, and substrate-binding mode in Acinetobacter baumannii MurF.
ATP-binding mode including a carbamoylated lysine and two Mg(2+) ions, and substrate-binding mode in Acinetobacter baumannii MurF.
Biochemical and biophysical research communications (2014-07-01)
Sun-Shin Cha, Young Jun An, Chang-Sook Jeong, Jeong Hee Yu, Kyung Min Chung
ZUSAMMENFASSUNG
MurF adds d-Ala-d-Ala dipeptide to UDP-N-acetylmuramyl-l-Ala-γ-d-Glu-m-DAP (or l-Lys) in an ATP-dependent manner, which is the last step in the biosynthesis of monomeric precursor of peptidoglycan. Here we report crystal structures of two MurF-ATP complexes: the MurF-ATP complex and the MurF-ATP-UDP complex. The ATP-binding mode revealed by the crystal structure of the MurF-ATP complex confirms the previous biochemical demonstration that a carbamoylated lysine and two Mg(2+) ions are required for enzyme activity of MurF. The UDP-MurF interactions observed in the crystal structure of the MurF-ATP-UDP complex depict the characteristic substrate-binding mode of MurF. The emergence and dissemination of multidrug-resistant Acinetobacter baumannii strains are great threats to public health. Therefore, the structural information on A. baumannii MurF as a validated target for drug discovery will provide a framework to develop antibacterial agents against multidrug-resistant A. baumannii infections as well as to understand the reaction mechanism of MurF.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Lysin, ≥98% (TLC)
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysin, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Mangan, powder, ≥99.9% trace metals basis
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Supelco
L-Lysin, analytical standard
Sigma-Aldrich
Mangan, powder, −325 mesh, ≥99% trace metals basis
Supelco
L-Lysin -monohydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mangan, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)
Lysin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Lysin -acetat (Salz), ≥98% (HPLC)
Supelco
L-Lysin -monohydrochlorid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysinacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Mangan, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mangan, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mangan, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Mangan, rod, 10mm, diameter 4.0mm, cast, 99.5%
Lysin-acetat, European Pharmacopoeia (EP) Reference Standard
Mangan, rod, 50mm, diameter 4.0mm, cast, 99.5%
Mangan, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%