Merck
Clinical trial evidence to advance the science of cholecystectomy.
JAMA (2014-07-10)
Joseph S Solomkin
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Supelco
Amoxicillin Trihydrat, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Amoxicillin, United States Pharmacopeia (USP) Reference Standard
Supelco
Amoxicillin Trihydrat, VETRANAL®, analytical standard
Amoxicillin Trihydrat, European Pharmacopoeia (EP) Reference Standard
Amoxicillin-Trihydrat für die Leistungsverifizierung, European Pharmacopoeia (EP) Reference Standard